Merck
Differential expression of pentraxin 3 in neutrophils.
Experimental and molecular pathology (2014-12-03)
Olga Razvina, Shuying Jiang, Koichi Matsubara, Riuko Ohashi, Go Hasegawa, Takashi Aoyama, Kenji Daigo, Tatsuhiko Kodama, Takao Hamakubo, Makoto Naito
ABSTRACT
Pentraxins belong to the superfamily of conserved proteins that are characterized by a cyclic multimeric structure. Pentraxin 3 (PTX3) is a long pentraxin which can be produced by different cell types upon exposure to various inflammatory signals. Inside the neutrophil PTX3 is stored in form of granules localized in the cytoplasm. Neutrophilic granules are divided into three types: azurophilic (primary) granules, specific (secondary) granules and gelatinase (tertiary) granules. PTX3 has been considered to be localized in specific (secondary) granules. Immunofluorescent analyses using confocal laser microscopic examination were performed to clarify the localization of all three groups of granules within the cytoplasm of the mature neutrophils and neutrophils stimulated with IL-8. Furthermore, PTX3 was localized in primary granules of promyelocyte cell line HL-60. As a result, we suggest that PTX3 is localized not only in specific granules, but is also partly expressed in primary and tertiary granules. After the stimulation with IL-8, irregular reticular structures called neutrophil extracellular traps (NETs) were formed, three types of granules were trapped by NETs and PTX3 showed partial colocalization with these granular components. PTX3 localized in all three types of granules in neutrophils may play important roles in host defense.
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Supelco
Dimethyl sulfoxide, analytical standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., dried, ≤0.02% water
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, puriss. p.a., ACS reagent, ≥99.9% (GC)
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Glutaraldehyde solution, technical, ~50% in H2O (5.6 M)
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use