Merck
  • An alternative to the classical α-arylation: the transfer of an intact 2-iodoaryl from ArI(O₂CCF₃)₂.
An alternative to the classical α-arylation: the transfer of an intact 2-iodoaryl from ArI(O₂CCF₃)₂.
Angewandte Chemie (International ed. in English) (2014-09-10)
Zhiyu Jia, Erik Gálvez, Rosa María Sebastián, Roser Pleixats, Ángel Álvarez-Larena, Eddy Martin, Adelina Vallribera, Alexandr Shafir
ABSTRACT
The α-arylation of carbonyl compounds is generally accomplished under basic conditions, both under metal catalysis and via aryl transfer from the diaryl λ(3)-iodanes. Here, we describe an alternative metal-free α-arylation using ArI(O2CCF3)2 as the source of a 2-iodoaryl group. The reaction is applicable to activated ketones, such as α-cyanoketones, and works with substituted aryliodanes. This formal C-H functionalization reaction is thought to proceed through a [3,3] rearrangement of an iodonium enolate. The final α-(2-iodoaryl)ketones are versatile synthetic building blocks.
Sigma-Aldrich
2-Acetylcyclohexanone, 97%
Sigma-Aldrich
[Bis(trifluoroacetoxy)iodo]benzene, purum, ≥95.0% (AT)
Sigma-Aldrich
[Bis(trifluoroacetoxy)iodo]benzene, 97%
Sigma-Aldrich
2-Isobutyrylcyclohexanone, 96%