Merck
  • Multisubstrate-compatible ELISA procedures for rapid and high-sensitivity immunoassays.
Multisubstrate-compatible ELISA procedures for rapid and high-sensitivity immunoassays.
Nature protocols (2011-03-18)
Chandra Kumar Dixit, Sandeep Kumar Vashist, Brian D MacCraith, Richard O'Kennedy
ABSTRACT
This protocol describes an improved and optimized approach to develop rapid and high-sensitivity ELISAs by covalently immobilizing antibody on chemically modified polymeric surfaces. The method involves initial surface activation with KOH and an O(2) plasma, and then amine functionalization with 3-aminopropyltriethoxysilane. The second step requires covalent antibody immobilization on the aminated surface, followed by ELISA. The ELISA procedure developed is 16-fold more sensitive than established methods. This protocol could be used generally as a quantitative analytical approach to perform high-sensitivity and rapid assays in clinical situations, and would provide a faster approach to screen phage-displayed libraries in antibody development facilities. The antibody immobilization procedure is of ∼3 h duration and facilitates rapid ELISAs. This method can be used to perform assays on a wide range of commercially relevant solid support matrices (including those that are chemically inert) with various biosensor formats.
Sigma-Aldrich
Peroxidase from horseradish, Type VI, essentially salt-free, lyophilized powder, ≥250 units/mg solid (using pyrogallol)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 2% 2-butanone, A15 MEK1, ≥99.8% (based on denaturant-free substance)
Sigma-Aldrich
Ethanol, purum, absolute ethanol, denaturated with 4.8% isopropanol, A15 IPA1, ≥99.8% (based on denaturant-free substance)