Merck
  • Filamin A expression correlates with proliferation and invasive properties of human metastatic melanoma tumors: implications for survival in patients.
Filamin A expression correlates with proliferation and invasive properties of human metastatic melanoma tumors: implications for survival in patients.
Journal of cancer research and clinical oncology (2014-06-09)
Kai Zhang, Tienian Zhu, Dongmei Gao, Yimei Zhang, Qinglan Zhao, Shuang Liu, Tongyi Su, Michel Bernier, Ruijing Zhao
ABSTRACT
Filamin A (FLNa) cross-links actin filaments into dynamic orthogonal networks and interacts with binding proteins of diverse cellular functions that are implicated in cell growth and motility regulation. Here, we tested the hypothesis that FLNa plays a role in cancer proliferation and metastasis via the regulation of epidermal growth factor receptor (EGFR) function. Ectopic expression and knockdown of FLNa in human melanoma cell lines was performed to investigate changes in cellular proliferation, migration and invasion in vitro and tumor growth in a xenograft model in the mouse. The role of FLNa in EGFR expression and signaling was evaluated by Western blot. Immunohistochemistry was performed on histological sections of human melanoma tumors to determine whether an association existed between FLNa and overall survival. The depletion of FLNa significantly reduced the proliferation, migration and invasion of two melanoma cell lines in vitro and was associated with smaller tumors in a xenograft model in vivo. EGF-induced phosphorylation of EGFR and activation of the Raf-MEK-ERK cascade was negatively affected by the silencing of FLNa both in vitro and in vivo. Cancer patients with low melanoma tumor FLNa expression have improved survival benefit. These data indicate that enhanced tumorigenesis occurs through increase in EGF-induced EGFR activation in FLNa-expressing melanoma cells and that high FLNa levels are predictors of negative outcome for patients with melanoma tumors.
Sigma-Aldrich
Anti-EGFR Antibody, neutralizing, clone LA1, clone LA1, Upstate®, from mouse
Sigma-Aldrich
O-Phospho-L-tyrosine
Sigma-Aldrich
Anti-FLNA antibody produced in rabbit, Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-Filamin A Antibody, clone TI10, ascites fluid, clone TI10, Chemicon®