Merck
  • Thermodynamics-based mathematical model for solubility prediction of glibenclamide in ethanol-water mixtures.
Thermodynamics-based mathematical model for solubility prediction of glibenclamide in ethanol-water mixtures.
Pharmaceutical development and technology (2013-08-13)
Faiyaz Shakeel, Fars K Alanazi, Ibrahim A Alsarra, Nazrul Haq
ABSTRACT
Temperature-dependent solubility data of glibenclamide (GBN) in various ethanol-water mixtures is not reported in literature so far. Therefore, the aim of this study was to determine the mole fraction solubility of GBN in various ethanol-water mixtures at the temperature range of 293.15 to 318.15 K. The solubility of GBN was determined by reported shake flask method and the experimental data was fitted in thermodynamics-based modified Apelblat model. The solubility of GBN was found to be increased with increase in temperature and mass fraction of ethanol in ethanol-water mixtures. The experimental data of GBN was well correlated with the modified Apelblat model at each temperature range with correlation coefficient of 0.9940-1.0000. The relative absolute deviation (AD) was found to be less than 0.1% except in pure ethanol and water. The positive values of enthalpies and entropies for GBN dissolution indicated that its dissolution is endothermic and an entropy-driven process.
Sigma-Aldrich
Glyburide, meets USP testing specifications
Sigma-Aldrich
Glybenclamide, ≥99% (HPLC)
Glybenclamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Glyburide (Glibenclamide), Pharmaceutical Secondary Standard; Certified Reference Material
Glibenclamide for peak identification, European Pharmacopoeia (EP) Reference Standard