Merck
  • Technical considerations on intra-arterial papaverine hydrochloride for cerebral vasospasm.
Technical considerations on intra-arterial papaverine hydrochloride for cerebral vasospasm.
Neuroradiology (1997-02-01)
J M Mathis, M E Jensen, J E Dion
ABSTRACT
Intra-arterial infusion of papaverine hydrochloride for subarachnoid hemorrhage-induced cerebral vasospasm has become an adjunctive endovascular therapy along with cerebral angioplasty. Our knowledge concerning the mechanism of action, method of administration and potential side effects of this therapeutic alternative are reviewed.
Product Description
Supelco
Papaverine hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Papaverine hydrochloride, powder
Papaverine hydrochloride, European Pharmacopoeia (EP) Reference Standard