Merck
  • Aplasia cutis congenita after exposure to methimazole: a causal relationship?
Aplasia cutis congenita after exposure to methimazole: a causal relationship?
The British journal of dermatology (1995-12-01)
T Vogt, W Stolz, M Landthaler
ABSTRACT
We report a child with scalp aplasia cutis congenita, whose mother was treated with methimazole during pregnancy. The relationship between antithyroid drug administration during pregnancy and the occurrence of scalp defects is discussed, and the pertinent literature is reviewed.
Product Description
Supelco
Methimazole, analytical standard
Supelco
Methimazole, VETRANAL®, analytical standard
Sigma-Aldrich
2-Mercapto-1-methylimidazole, ≥99%
USP
Methimazole, United States Pharmacopeia (USP) Reference Standard
Thiamazole, European Pharmacopoeia (EP) Reference Standard