Merck
Fragrance material review on linalool.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2003-06-14)
C S Letizia, J Cocchiara, J Lalko, A M Api
ABSTRACT
A toxicologic and dermatologic review of linalool when used as a fragrance ingredient, is presented.
Sigma-Aldrich
L-Linalool, natural, ≥95%, FG
Sigma-Aldrich
(−)-Linalool, ≥95.0% (sum of enantiomers, GC)
Supelco
(−)-Linalool, analytical standard
Sigma-Aldrich
Linalool, ≥97%, FCC, FG
Sigma-Aldrich
Linalool, 97%
Supelco
Linalool, analytical standard