Merck
  • Specification of spatial identities of cerebellar neuron progenitors by ptf1a and atoh1 for proper production of GABAergic and glutamatergic neurons.
Specification of spatial identities of cerebellar neuron progenitors by ptf1a and atoh1 for proper production of GABAergic and glutamatergic neurons.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2014-04-04)
Mayumi Yamada, Yusuke Seto, Shinichiro Taya, Tomoo Owa, Yukiko U Inoue, Takayoshi Inoue, Yoshiya Kawaguchi, Yo-Ichi Nabeshima, Mikio Hoshino
ABSTRACT
In the cerebellum, the bHLH transcription factors Ptf1a and Atoh1 are expressed in distinct neuroepithelial regions, the ventricular zone (VZ) and the rhombic lip (RL), and are required for producing GABAergic and glutamatergic neurons, respectively. However, it is unclear whether Ptf1a or Atoh1 is sufficient for specifying GABAergic or glutamatergic neuronal fates. To test this, we generated two novel knock-in mouse lines, Ptf1a(Atoh1) and Atoh1(Ptf1a), that are designed to express Atoh1 and Ptf1a ectopically in the VZ and RL, respectively. In Ptf1a(Atoh1) embryos, ectopically Atoh1-expressing VZ cells produced glutamatergic neurons, including granule cells and deep cerebellar nuclei neurons. Correspondingly, in Atoh1(Ptf1a) animals, ectopically Ptf1a-expressing RL cells produced GABAergic populations, such as Purkinje cells and GABAergic interneurons. Consistent results were also obtained from in utero electroporation of Ptf1a or Atoh1 into embryonic cerebella, suggesting that Ptf1a and Atoh1 are essential and sufficient for GABAergic versus glutamatergic specification in the neuroepithelium. Furthermore, birthdating analyses with BrdU in the knock-in mice or with electroporation studies showed that ectopically produced fate-changed neuronal types were generated at temporal schedules closely simulating those of the wild-type RL and VZ, suggesting that the VZ and RL share common temporal information. Observations of knock-in brains as well as electroporated brains revealed that Ptf1a and Atoh1 mutually negatively regulate their expression, probably contributing to formation of non-overlapping neuroepithelial domains. These findings suggest that Ptf1a and Atoh1 specify spatial identities of cerebellar neuron progenitors in the neuroepithelium, leading to appropriate production of GABAergic and glutamatergic neurons, respectively.
Sigma-Aldrich
L-Glutamic acid hydrochloride
Sigma-Aldrich
γ-Aminobutyric acid, ≥99%
Glutamic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
γ-Aminobutyric acid, analytical standard
Vigabatrin impurity D, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
γ-Aminobutyric acid, BioXtra, ≥99%
Supelco
L-Glutamic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
D-Glutamic acid, ≥99% (TLC)
Sigma-Aldrich
L-Glutamic acid, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-100.5%
Sigma-Aldrich
L-Glutamic acid, ReagentPlus®, ≥99% (HPLC)
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Glutamic acid, BioUltra, ≥99.5% (NT)