Merck
  • Intestine-specific deletion of SIRT1 in mice impairs DCoH2-HNF-1α-FXR signaling and alters systemic bile acid homeostasis.
Intestine-specific deletion of SIRT1 in mice impairs DCoH2-HNF-1α-FXR signaling and alters systemic bile acid homeostasis.
Gastroenterology (2014-01-07)
Nevzat Kazgan, Mallikarjuna R Metukuri, Aparna Purushotham, Jing Lu, Anuradha Rao, Sangkyu Lee, Matthew Pratt-Hyatt, Andrew Lickteig, Iván L Csanaky, Yingming Zhao, Paul A Dawson, Xiaoling Li
ABSTRACT
Sirtuin 1 (SIRT1), the most conserved mammalian oxidized nicotinamide adenine dinucleotide-dependent protein deacetylase, is an important metabolic sensor in many tissues. However, little is known about its role in the small intestine, which absorbs and senses nutrients. We investigated the functions of intestinal SIRT1 in systemic bile acid and cholesterol metabolism in mice. SIRT1 was specifically deleted from the intestines of mice using the flox-Villin-Cre system (SIRT1 iKO mice). Intestinal and hepatic tissues were collected, and bile acid absorption was analyzed using the everted gut sac experiment. Systemic bile acid metabolism was studied in SIRT1 iKO and flox control mice placed on standard diets, diets containing 0.5% cholic acid or 1.25% cholesterol, or lithogenic diets. SIRT1 iKO mice had reduced intestinal farnesoid X receptor (FXR) signaling via hepatocyte nuclear factor 1α (HNF-1α) compared with controls, which reduced expression of the bile acid transporter genes Asbt and Mcf2l (encodes Ost) and absorption of ileal bile acids. SIRT1 regulated HNF-1α/FXR signaling partially through dimerization cofactor of HNF-1a (Dcoh2) Dcoh2, which increases dimerization of HNF-1α. SIRT1 was found to deacetylate Dcoh2, promoting its interaction with HNF-1α and inducing DNA binding by HNF-1α. Intestine-specific deletion of SIRT1 increased hepatic bile acid biosynthesis, reduced hepatic accumulation of bile acids, and protected animals from liver damage from a diet high in levels of bile acids. Intestinal SIRT1, a key nutrient sensor, is required for ileal bile acid absorption and systemic bile acid homeostasis in mice. We delineated the mechanism of metabolic regulation of HNF-1α/FXR signaling. Reagents designed to inhibit intestinal SIRT1 might be developed to treat bile acid-related diseases such as cholestasis.
Product Description
Cholic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%