Merck
  • Monolignol ferulate transferase introduces chemically labile linkages into the lignin backbone.
Monolignol ferulate transferase introduces chemically labile linkages into the lignin backbone.
Science (New York, N.Y.) (2014-04-05)
C G Wilkerson, S D Mansfield, F Lu, S Withers, J-Y Park, S D Karlen, E Gonzales-Vigil, D Padmakshan, F Unda, J Rencoret, J Ralph
ABSTRACT
Redesigning lignin, the aromatic polymer fortifying plant cell walls, to be more amenable to chemical depolymerization can lower the energy required for industrial processing. We have engineered poplar trees to introduce ester linkages into the lignin polymer backbone by augmenting the monomer pool with monolignol ferulate conjugates. Herein, we describe the isolation of a transferase gene capable of forming these conjugates and its xylem-specific introduction into poplar. Enzyme kinetics, in planta expression, lignin structural analysis, and improved cell wall digestibility after mild alkaline pretreatment demonstrate that these trees produce the monolignol ferulate conjugates, export them to the wall, and use them during lignification. Tailoring plants to use such conjugates during cell wall biosynthesis is a promising way to produce plants that are designed for deconstruction.
Sigma-Aldrich
trans-Ferulic acid, ≥99%
Sigma-Aldrich
trans-Ferulic acid, 99%
Supelco
trans-Ferulic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Ferulic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
4-Hydroxy-3-methoxycinnamic acid, mixture of isomers, analytical standard
Supelco
trans-Ferulic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)