Merck
  • Identification of pre-leukaemic haematopoietic stem cells in acute leukaemia.
Identification of pre-leukaemic haematopoietic stem cells in acute leukaemia.
Nature (2014-02-14)
Liran I Shlush, Sasan Zandi, Amanda Mitchell, Weihsu Claire Chen, Joseph M Brandwein, Vikas Gupta, James A Kennedy, Aaron D Schimmer, Andre C Schuh, Karen W Yee, Jessica L McLeod, Monica Doedens, Jessie J F Medeiros, Rene Marke, Hyeoung Joon Kim, Kwon Lee, John D McPherson, Thomas J Hudson, Andrew M K Brown, Fouad Yousif, Quang M Trinh, Lincoln D Stein, Mark D Minden, Jean C Y Wang, John E Dick
ABSTRACT
In acute myeloid leukaemia (AML), the cell of origin, nature and biological consequences of initiating lesions, and order of subsequent mutations remain poorly understood, as AML is typically diagnosed without observation of a pre-leukaemic phase. Here, highly purified haematopoietic stem cells (HSCs), progenitor and mature cell fractions from the blood of AML patients were found to contain recurrent DNMT3A mutations (DNMT3A(mut)) at high allele frequency, but without coincident NPM1 mutations (NPM1c) present in AML blasts. DNMT3A(mut)-bearing HSCs showed a multilineage repopulation advantage over non-mutated HSCs in xenografts, establishing their identity as pre-leukaemic HSCs. Pre-leukaemic HSCs were found in remission samples, indicating that they survive chemotherapy. Therefore DNMT3A(mut) arises early in AML evolution, probably in HSCs, leading to a clonally expanded pool of pre-leukaemic HSCs from which AML evolves. Our findings provide a paradigm for the detection and treatment of pre-leukaemic clones before the acquisition of additional genetic lesions engenders greater therapeutic resistance.
Sigma-Aldrich
Isocitrate Dehydrogenase 1 (NADP+) human, recombinant, expressed in Sf9 cells, ≥90% (SDS-PAGE)
Sigma-Aldrich
Isocitrate Dehydrogenase 1 (NADP+) human, recombinant, expressed in E. coli, lyophilized powder, ≥80 units/mg protein
Sigma-Aldrich
Isocitric Dehydrogenase (NADP) from porcine heart, Type IV, buffered aqueous glycerol solution, 3-20 units/mg protein
Sigma-Aldrich
Isocitric Dehydrogenase (NADP) from porcine heart, Type I, 0.5-3.0 unit/mg solid (plus numerous enzyme activities associated with porcine heart)