Merck
  • Effect of saliva stimulants (Hybrin and malic acid) on cervical root surfaces in vitro.
Effect of saliva stimulants (Hybrin and malic acid) on cervical root surfaces in vitro.
Scandinavian journal of dental research (1980-06-01)
G Anneroth, G Nordenram, S Bengtsson
ABSTRACT
Two saliva stimulating drugs, solutions of Hybrin and malic acid were in an in vitro study found to have a heavy demineralizing effect on the root surface of the dental hard tissue. Consequently the drugs cannot be recommended to patients suffering from dry mouth reaction.
Sigma-Aldrich
Malic acid, meets USP/NF testing specifications
Sigma-Aldrich
DL-Malic acid, ≥98% (capillary GC)
Sigma-Aldrich
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
Sigma-Aldrich
DL-Malic acid, 99%
Sigma-Aldrich
DL-Malic acid, ReagentPlus®, ≥99%