Merck
  • [Characterization of puroindolines in the control of endosperm texture in common wheat lines with substitutions of homeologous group-5 chromosomes].
[Characterization of puroindolines in the control of endosperm texture in common wheat lines with substitutions of homeologous group-5 chromosomes].
Genetika (2013-06-13)
L V Obukhova, T T Efremova, V K Shumnyĭ
ABSTRACT
The genetic control of grain hardness and its association with the specific friabilin content on starch granules of common wheat cultivars and lines with intervarietal substitutions of homeologous group-5 chromosomes were studied. A significant correlation was revealed between the technological parameters of grain hardness (mean size of flour particles) and the specific content of puroindolines on the starch surface estimated in terms of starch doses. The results obtained allowed the method of starch doses to be used to identify soft and hard wheat cultivars and lines based on an analysis of a single grain. The biochemical analysis confirmed the previously obtained estimates of flour-grinding properties of wheat cultivars and substitution lines, which allowed specific genotypes to be characterized according to the composition of puroindolines. The influence of chromosomes 5D and 5A of donor wheat cultivars on the activity of the Ha loci of recipient cultivars was revealed and found to be associated with the composition of PIN products and with the expression of the Pina-D1 and Pinb-D1 genes.
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch from rice
Sigma-Aldrich
Starch from corn
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from corn, practical grade
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.