Merck
  • Intake of caffeinated, carbonated, or citrus beverage types and development of lower urinary tract symptoms in men and women.
Intake of caffeinated, carbonated, or citrus beverage types and development of lower urinary tract symptoms in men and women.
American journal of epidemiology (2013-06-01)
Nancy N Maserejian, Carrie G Wager, Edward L Giovannucci, Teresa M Curto, Kevin T McVary, John B McKinlay
ABSTRACT
Evidence to substantiate recommendations for restriction of caffeinated or acidic beverages as self-management for lower urinary tract symptoms (LUTS) is limited. We examined longitudinal and acute associations between beverage intake and LUTS in the Boston Area Community Health (BACH) cohort (n = 4,144) between 2002 and 2010. Multivariable models tested associations between baseline intakes and progression of LUTS at 5-year follow-up, between follow-up intakes and International Prostate Symptom Scores at follow-up, and between 5-year intake changes and LUTS progression. Greater coffee or total caffeine intake at baseline increased the odds of LUTS progression in men (coffee: >2 cups/day vs. none, odds ratio = 2.09, 95% confidence interval: 1.29, 3.40, P-trend = 0.01; caffeine: P-trend < 0.001), particularly storage symptoms. Women who increased coffee intake by at least 2 servings/day during follow-up (compared with categories of decreased or unchanged intakes) had 64% higher odds of progression of urgency (P = 0.003). Women with recently increased soda intake, particularly caffeinated diet soda, had higher symptom scores, urgency, and LUTS progression. Citrus juice intake was associated with 50% lower odds of LUTS progression in men (P = 0.02). Findings support recommendations to limit caffeinated beverage intake for LUTS, and in men, they suggest benefits of citrus juice consumption. Further clinical research is warranted, particularly of the precise role of sodas containing artificial sweeteners in bladder sensations and urological function.
Product Description
Supelco
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Caffeine, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Caffeine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Melting point standard 235-237°C, analytical standard