Merck
  • Chitosan based advanced hemostatic dressing is associated with decreased blood loss in a swine uncontrolled hemorrhage model.
Chitosan based advanced hemostatic dressing is associated with decreased blood loss in a swine uncontrolled hemorrhage model.
American journal of surgery (2013-03-19)
Nicholas R Kunio, Gordon M Riha, Katherine M Watson, Jerome A Differding, Martin A Schreiber, Jennifer M Watters
ABSTRACT
The purpose of this study was to compare standard gauze (SG) and advanced hemostatic dressings in use by military personnel in a no-hold model. A randomized, controlled trial was conducted using 36 swine. Animals underwent femoral arteriotomy, followed by 60 seconds of uncontrolled hemorrhage. After hemorrhage, packing with 1 of 3 dressings-SG, Combat Gauze (CG), or Celox Rapid gauze (XG)-and a 500-mL bolus of Hextend were initiated. Pressure was not held after packing, and animals were followed for 120 minutes. Physiologic parameters were monitored continuously, and electrolyte and hematologic laboratory assessments were performed before injury and 30 and 120 minutes after injury. Dressing failure was determined if bleeding occurred outside the wound. All animals survived to study end. Baseline characteristics were similar between groups. No statistical difference was seen in initial blood loss or dressing success rate (SG, 10 of 12; CG, 10 of 12; and XG, 12 of 12). Secondary blood loss was significantly less with XG (median, 12.8 mL; interquartile range, 8.8 to 39.7 mL) compared with SG (median, 44.7 mL; interquartile range, 17.8 to 85.3 mL; P = .02) and CG (median, 31.9 mL; interquartile range, 18.6 to 69.1 mL; P = .05). Packing time was significantly shorter with XG (mean, 37.1 ± 6.2 seconds) compared with SG (mean, 45.2 ± 6.0 seconds; P < .01) and CG (mean, 43.5 ± 5.6 seconds; P = .01). XG demonstrated shorter application time and decreased secondary blood loss in comparison with both SG and CG. These differences may be of potential benefit in a care-under-fire scenario.
Sigma-Aldrich
Kaolin, heavy, powder
Sigma-Aldrich
Kaolin
Sigma-Aldrich
Kaolin, tested according to Ph. Eur.
Sigma-Aldrich
Kaolin, meets USP testing specifications
Sigma-Aldrich
Halloysite nanoclay