Merck
  • Mechanism of Hsp104/ClpB inhibition by prion curing Guanidinium hydrochloride.
Mechanism of Hsp104/ClpB inhibition by prion curing Guanidinium hydrochloride.
FEBS letters (2013-02-19)
Eva Kummer, Yuki Oguchi, Fabian Seyffer, Bernd Bukau, Axel Mogk
ABSTRACT
The Saccharomyces cerevisiae AAA+ protein Hsp104 and its Escherichia coli counterpart ClpB cooperate with Hsp70 chaperones to refold aggregated proteins and fragment prion fibrils. Hsp104/ClpB activity is regulated by interaction of the M-domain with the first ATPase domain (AAA-1), controlling ATP turnover and Hsp70 cooperation. Guanidinium hydrochloride (GdnHCl) inhibits Hsp104/ClpB activity, leading to prion curing. We show that GdnHCl binding exerts dual effects on Hsp104/ClpB. First, GdnHCl strengthens M-domain/AAA-1 interaction, stabilizing Hsp104/ClpB in a repressed conformation and abrogating Hsp70 cooperation. Second, GdnHCl inhibits continuous ATP turnover by AAA-1. These findings provide the mechanistic basis for prion curing by GdnHCl.
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Guanidine sulfate salt, 99%
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)