Merck
[The rare case of Bland-White-Garland syndrome in adult patient].
Kardiologiia (2012-12-15)
Iu M Alekseeva, V I Potievskaia, E A Sakovich, A V Sitnikov, E L Soiustova, V A Fin'ko, A V Ardashev
ABSTRACT
We present analysis of a case history of a patient with rare congenital heart disorder - Bland-White-Garland syndrome. The 25 years old women was first diagnosed with this disorder during an examination in the cardiological department of the Moscow clinical hospital No83. The paper contains discussion of difficulties of diagnosis and peculiarities of management of adults with this pathology as well as of a problem of the choice of further therapeutic approaches.
Sigma-Aldrich
(±)-Metoprolol (+)-tartrate salt, ≥98% (titration), powder
Supelco
Metoprolol tartrate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Metoprolol tartrate, European Pharmacopoeia (EP) Reference Standard
Supelco
Metoprolol Tartrate, Pharmaceutical Secondary Standard; Certified Reference Material