Merck
  • Epirubicin loaded with propylene glycol liposomes significantly overcomes multidrug resistance in breast cancer.
Epirubicin loaded with propylene glycol liposomes significantly overcomes multidrug resistance in breast cancer.
Cancer letters (2012-11-29)
Ying-Zheng Zhao, Dan-Dan Dai, Cui-Tao Lu, Li-Juan Chen, Min Lin, Xiao-Tong Shen, Xiao-Kun Li, Ming Zhang, Xi Jiang, Rong-Rong Jin, Xing Li, Hai-Feng Lv, Lu Cai, Pin-Tong Huang
ABSTRACT
Multidrug resistance (MDR) is one of the major reasons for the failure of cancer chemotherapy. A newly reported liposome carrier, propylene glycol liposomes (EPI-PG-liposomes) were made to load epirubicin (EPI) which enhanced EPI absorption in MDR tumor cells to overcome the drug resistance. MDA-MB 435 and their mutant resistant (MDA-MB 435/ADR) cells were used to examine the cellular uptake and P-gp function in vitro for EPI-PG-liposomes by fluorescence microscopy and FCM, respectively. Mammary tumor model was also established to investigate the tumor growth inhibition and pharmacodynamics of EPI-PG-liposomes in vivo. Morphology evaluation showed that EPI-PG-liposomes had a homogeneous spherical shape with an average diameter of 182 nm. Based on cell viability assay, fluorescent microscopy examination, and EPI uptake assay, EPI-PG-liposomes exhibited an effective growth inhibition not only in MDA-MB-435 cells, but also in MDA-MB 435/ADR cells. EPI-PG-liposomes have high permeability not only on tumor cell membrane, but also on cell nucleus membrane. P-gp function assay showed that the anticancer action of EPI-PG-liposomes was not related to P-gp efflux pump, suggesting that PG-liposomes would not affect the normal physiological functions of membrane proteins. EPI-PG-liposomes also showed a better antitumor efficacy compared to EPI solution alone. With high entrapment efficiency, spherical morphology and effective inhibition on MDR cancer cells, EPI-PG-liposomes may represent a better chemotherapeutic vectors for cancer targeted therapy.
Sigma-Aldrich
(S)-(+)-1,2-Propanediol, 96%
Sigma-Aldrich
1,2-Propanediol, tested according to Ph. Eur.
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Supelco
1,2-Propanediol, analytical standard
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Supelco
Propylene glycol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(R)-(−)-1,2-Propanediol, 96%
Sigma-Aldrich
Propylene Glycol, meets USP testing specifications
Sigma-Aldrich
Propylene glycol, ≥99.5% (GC), FCC, FG