Merck
  • Chiroptical sensing of citronellal: systematic development of a stereodynamic probe using the concept of isostericity.
Chiroptical sensing of citronellal: systematic development of a stereodynamic probe using the concept of isostericity.
Chemical communications (Cambridge, England) (2012-10-16)
Daniel P Iwaniuk, Christian Wolf
ABSTRACT
A rationally designed stereodynamic ICD probe carrying two terminal amino functions for chemo- and enantioselective sensing of citronellal is introduced. The sensor shows strong Cotton effects upon diimine formation which can be used for accurate ee determination.
Sigma-Aldrich
(R)-(+)-Citronellal, technical grade
Supelco
(±)-Citronellal, analytical standard
Sigma-Aldrich
(±)-Citronellal, natural, ≥85%, FCC, FG
Sigma-Aldrich
(S)-(−)-Citronellal, 96%
Sigma-Aldrich
(±)-Citronellal, ≥95.0% (GC)