Merck
  • I2-CF3SO3H synergistic promoted sp3 C-H bond diarylation of aromatic ketones.
I2-CF3SO3H synergistic promoted sp3 C-H bond diarylation of aromatic ketones.
Organic letters (2012-10-13)
Yan-ping Zhu, Feng-cheng Jia, Mei-cai Liu, Liu-ming Wu, Qun Cai, Yang Gao, An-xin Wu
ABSTRACT
An I(2)-CF(3)SO(3)H synergistic promoted sp(3) C-H bond diarylation protocol was developed for the synthesis of 2,2-bis(4-(dimethylamino)phenyl)-1-aryl ethanones. The reaction performed well in the absence of any metal and ligand. It integrated three reactions with different mechanisms (iodination, Kornblum oxidation, and hydroarylation) in a single reactor.
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 96%
Sigma-Aldrich
Trifluoromethanesulfonic acid, reagent grade, 98%
Sigma-Aldrich
Lanthanum(III) trifluoromethanesulfonate, 99.999% trace metals basis
Sigma-Aldrich
Lithium trifluoromethanesulfonate, 99.995% trace metals basis
Sigma-Aldrich
Trifluoromethanesulfonic acid, ReagentPlus®, ≥99%
Sigma-Aldrich
Zinc trifluoromethanesulfonate, 98%
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99%
Sigma-Aldrich
Aluminum trifluoromethanesulfonate, 99.9% trace metals basis
Sigma-Aldrich
Silver trifluoromethanesulfonate, purum, ≥98.0% (Ag)
Sigma-Aldrich
Silver trifluoromethanesulfonate, ≥99.95% trace metals basis