Merck
  • Removal of anionic dyes from aqueous solutions by an ion-exchanger based on pullulan microspheres.
Removal of anionic dyes from aqueous solutions by an ion-exchanger based on pullulan microspheres.
Carbohydrate polymers (2012-10-10)
Marieta Constantin, Ionela Asmarandei, Valeria Harabagiu, Luminita Ghimici, Paolo Ascenzi, Gheorghe Fundueanu
ABSTRACT
Pullulan-graft-poly(3-acrylamidopropyl trimethylammonium chloride) (P-g-pAPTAC) microspheres were prepared by suspension cross-linking of the pullulan previously grafted with cationic moieties. Adsorption of Azocarmine B by the P-g-pAPTAC microspheres was used as a model to demonstrate the removal of anionic dyes from aqueous solutions. Batch adsorption studies concerning the effect of the contact time, pH, initial dye concentration, temperature, grafting, and the nature of sulfonated anionic dyes on the adsorption kinetics were investigated. Adsorption was shown to be independent of pH. The experimental data best fitted to the pseudo-second order model which provided values of the rate constant k(2) of 1.4×10(-4) g mg(-1) min(-1) for 100 mg L(-1) solution and of 3.7×10(-4) g mg(-1) min(-1) for 500 mg L(-1) solution. From the Langmuir isotherm linear equation, the maximum adsorption capacity determined was 113.63 mg of Azocarmine B per gram of adsorbent; the negative value of the free energy change indicated the spontaneous nature of the adsorption process.
Sigma-Aldrich
Pullulan, suitable for substrate for pullulanase
Supelco
Pullulan Standard Set Mp ~350 - 700′000, set of analytical standards