Merck
  • Two-step pretreatment of corn stalk silage for increasing sugars production and decreasing the amount of catalyst.
Two-step pretreatment of corn stalk silage for increasing sugars production and decreasing the amount of catalyst.
Bioresource technology (2012-07-10)
Runjuan Wang, Youshan Sun, Shuting Zhang, Xuebin Lu
ABSTRACT
The study investigated the effects of two-step pretreatment on fermentable sugar production from corn stalk silage. In the first step, the corn stalk silage was extracted by tepid water and then the solid was pretreated using Fe(NO(3))(3) as catalyst. The results showed that 45.8 g/100g DM total sugars was obtained and the surface of remaining solid was seriously damaged after two-step pretreatment. Compared with one-step pretreatment, the production of total sugars increased by 23.8% and the amount of the catalyst of Fe(NO(3))(3) decreased by 28.8%. This research provides a new effective, suitable and economical pretreatment method for biogas production from corn stalk.
Sigma-Aldrich
Iron(III) nitrate nonahydrate, ≥99.95% trace metals basis
Sigma-Aldrich
Iron(III) nitrate nonahydrate, ≥99.999% trace metals basis
Sigma-Aldrich
Iron(III) nitrate nonahydrate, BioReagent, suitable for cell culture
Sigma-Aldrich
Iron(III) nitrate nonahydrate, ACS reagent, ≥98%