Merck
Biodegradation of triclosan by a wastewater microorganism.
Water research (2012-06-08)
Do Gyun Lee, Fuman Zhao, Yohannes H Rezenom, David H Russell, Kung-Hui Chu
ABSTRACT
Triclosan, a synthetic antimicrobial agent, has been considered as an emerging environmental contaminant. Here we reported a triclosan-degrading wastewater bacterial isolate, Sphingopyxis strain KCY1, capable of dechlorinating triclosan with a stoichiometric release of chloride. The stain can degrade diphenyl ether but not 2,4,4'-tribromodiphenyl ether and 2,2',4,4'-tetrabromodiphenyl ether, despite all these three compounds are structurally similar to triclosan. While strain KCY1 was unable to grow on triclosan and catechol, it could grow with glucose, sodium succinate, sodium acetate, and phenol. When grown with complex nutrient medium containing a trace amount of triclosan (as low as 5 μg/L), the strain could retain its degradation ability toward triclosan. The maximum-specific triclosan degradation rate (q(m)) and the half-velocity constant (K(m)) are 0.13 mg-triclosan/mg-protein/day and 2.8 mg-triclosan/L, respectively. As triclosan degradation progressed, five metabolites were identified and these metabolites continue to transform into non-chlorinated end products, which was supported by a sharp drop in androgenic potential. The activity of catechol 2,3-dioxygenase in the cell extract was detected. No triclosan degradation was observed in the presence of 3-fluorocatechol, an inhibitor of meta-cleavage enzyme, suggesting that triclosan degradation proceed via meta-cleavage pathway. Based on all the observations, a degradation pathway for triclosan by strain KCY1 was proposed.
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Supelco
Diphenyl ether, Selectophore, ≥99.9%
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, ≥99%
Sigma-Aldrich
Diphenyl ether, ReagentPlus®, 99%
Sigma-Aldrich
Diphenyl ether, ≥99%, FG
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
Sigma-Aldrich
Sodium acetate trihydrate, ACS reagent, ≥99%