Merck
  • Synthesis and evaluation of atropos dihydro-5H-dibenzazepinium halide PTCs derived from α-methylbenzylamine.
Synthesis and evaluation of atropos dihydro-5H-dibenzazepinium halide PTCs derived from α-methylbenzylamine.
Organic & biomolecular chemistry (2012-05-23)
Barry Lygo, Umar Butt, Maria Cormack
ABSTRACT
A short synthetic route to diastereoisomeric atropos dihydro-5H-dibenz[c,e]azepinium salts via reaction of a single enantiomer of (R)-α-methylbenzylamine with a racemic atropos biphenol derivative is described. Compounds prepared via this approach are used to provide strong evidence that structurally related tropos dihydro-5H-dibenz[c,e]azepinium salts preferentially react via a single conformation in PTC reactions involving glycine imine enolates.
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, ChiPros®, produced by BASF, ≥99.0%
Supelco
(R)-(+)-α-Methylbenzylamine, for chiral derivatization, LiChropur, ≥99.0%
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, purum, ≥98.0% (sum of enantiomers, GC)
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, 98%
Sigma-Aldrich
α-Methylbenzylamine, 99%
Sigma-Aldrich
(S)-(−)-α-Methylbenzylamine, 98%