- Metal-free nitrogen-containing carbon nanotubes prepared from triazole and tetrazole derivatives show high electrocatalytic activity towards the oxygen reduction reaction in alkaline media.
Metal-free nitrogen-containing carbon nanotubes prepared from triazole and tetrazole derivatives show high electrocatalytic activity towards the oxygen reduction reaction in alkaline media.
ChemSusChem (2012-03-06)
Adina Morozan, Pascale Jégou, Mathieu Pinault, Stéphane Campidelli, Bruno Jousselme, Serge Palacin
PMID22389330
ABSTRACT
High-performance oxygen reduction reaction (ORR) catalysts based on metal-free nitrogen-containing precursors and carbon nanotubes are reported. The investigated systems allow the evaluation of the effect of nitrogen-containing groups towards ORR and the results show that the catalysts are compatible with the conditions encountered in alkaline fuel cells, exhibiting good catalytic activity and stability compared with conventional Pt/C electrocatalyst.
Sigma-Aldrich
Tetrazole solution, suitable for DNA synthesis, filtered through a 1 μm filter, ~0.45 M in acetonitrile