Merck
  • Osteoconductivity of thermal-sprayed silver-containing hydroxyapatite coating in the rat tibia.
Osteoconductivity of thermal-sprayed silver-containing hydroxyapatite coating in the rat tibia.
The Journal of bone and joint surgery. British volume (2011-04-23)
Y Yonekura, H Miyamoto, T Shimazaki, Y Ando, I Noda, M Mawatari, T Hotokebuchi
ABSTRACT
A silver-containing hydroxyapatite (Ag-HA) coating has been developed using thermal spraying technology. We evaluated the osteoconductivity of this coating on titanium (Ti) implants in rat tibiae in relation to bacterial infection in joint replacement. At 12 weeks, the mean affinity indices of bone formation of a Ti, an HA, a 3%Ag-HA and a 50%Ag-HA coating were 97.3%, 84.9%, 81.0% and 40.5%, respectively. The mean affinity indices of bone contact of these four coatings were 18.8%, 83.7%, 77.2% and 40.5%, respectively. The indices of bone formation and bone contact around the implant of the 3%Ag-HA coating were similar to those of the HA coating, and no significant differences were found between them (bone formation, p = 0.99; bone contact, p = 0.957). However, inhibition of bone formation was observed with the 50%Ag-HA coating. These results indicate that the 3%Ag-HA coating has low toxicity and good osteoconductivity, and that the effect of silver toxicity on osteoconductivity depends on the dose.
Sigma-Aldrich
Silver oxide, predominantly silver(II) oxide
Sigma-Aldrich
Silver(I) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Silver(I) oxide, ReagentPlus®, 99%
Sigma-Aldrich
Silver(I) oxide, purum p.a., ≥99.0% (AT)