Merck
Extracorporeal life support in a severe Taxus baccata poisoning.
Clinical toxicology (Philadelphia, Pa.) (2010-06-10)
Claudia Panzeri, Giuseppe Bacis, Francesco Ferri, Gabriele Rinaldi, Andrea Persico, Francesca Uberti, Patrizia Restani
ABSTRACT
Yew (Taxus baccata) is a conifer known to be toxic since ancient times. Taxine A and taxine B, the toxic alkaloids of Taxus, block cardiac sodium and calcium channels causing nausea, vomiting, abdominal pain, cardiac arrhythmias, respiratory distress, coma, seizures, and death in yew poisoning. A 44-year-old male farmer was admitted to the hospital because of a suspected myocardial infarction. First bradycardia and then ventricular tachycardia were present and a severe right ventricular dilatation with biventricular dysfunction was observed but with normal coronary arteriography. He was resistant to conventional therapy and, 6 h after hospital admission, extracorporeal support with membrane oxygenation was applied. The patient recovered. Nine days later, a large number of yew leaves were unexpectedly observed in his feces. Botanical and laboratory analysis confirmed the poisoning. Blood (651 ng/mL) and urinary (5.6 mcg/mL) levels of 3,5-dimethoxyphenol (metabolite of taxicatine) were greater than previously reported in lethal cases. The patient was transferred to a psychiatric unit 17 days after admission. Intensive treatment of severe cardiovascular symptoms with antiarrhythmic drugs, temporary pacemaker, intra-aortic balloon pump, extracorporeal membrane oxygenation, and extracorporeal life support can be life-saving even after a potentially lethal ingestion of T. baccata leaves.
Sigma-Aldrich
3,5-Dimethoxyphenol, 99%