Merck
  • Assessment of the extent of starch dissolution in dimethyl sulfoxide by 1H NMR spectroscopy.
Assessment of the extent of starch dissolution in dimethyl sulfoxide by 1H NMR spectroscopy.
Macromolecular bioscience (2008-12-18)
Sarah Schmitz, Anthony C Dona, Patrice Castignolles, Robert G Gilbert, Marianne Gaborieau
ABSTRACT
Complete dissolution is needed for the separation, characterization, or homogeneous labeling of whole starch molecules. A method is presented to quantify the extent of starch dissolution in DMSO for the first time; it is validated on a commercial rice starch. It is used directly on starch dispersions containing possible undissolved or co-dissolved species. High-amylose maize starches, known to be digested slowly in vivo, only quantitatively dissolve in the presence of high concentrations of an H-bond disrupter, LiBr, although they form clear dispersions at low LiBr concentrations. Starch quantitatively dissolves from waxy rice flours; non-starch components partially co-dissolve but do not interfere with the dissolution quantification.
Sigma-Aldrich
Lithium bromide solution, 54 wt. % in H2O
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Lithium bromide, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Lithium bromide, ReagentPlus®, ≥99%
Sigma-Aldrich
Lithium bromide, powder and chunks, ≥99.995% trace metals basis