Merck
  • Organocatalytic enantioselective Diels-Alder reaction of dienes with alpha-(N,N-Diacylamino)acroleins.
Organocatalytic enantioselective Diels-Alder reaction of dienes with alpha-(N,N-Diacylamino)acroleins.
Organic letters (2008-06-07)
Kazuaki Ishihara, Kazuhiko Nakano, Matsujiro Akakura
ABSTRACT
Catalytic and highly enantioselective Diels-Alder reaction of cyclic and acyclic dienes with alpha-phthalimidoacroleins provides cyclic alpha-quaternary alpha-amino acid precursors. The conformationally flexible chiral ammonium salt of H-L-Phe-L-Leu-N(CH(2)CH(2)) 2-reduced triamine with pentafluorobenzensulfonic acid is very effective as an asymmetric catalyst for the Diels-Alder reaction.
Sigma-Aldrich
Phthalimide potassium salt, purum, ≥99.0% (NT)
Sigma-Aldrich
Phthalimide potassium salt, 98%
Sigma-Aldrich
Phthalimide, ≥99%