Merck
  • Improved and efficient synthesis of chiral N,P-ligands via cyclic sulfamidates for asymmetric addition of butyllithium to benzaldehyde.
Improved and efficient synthesis of chiral N,P-ligands via cyclic sulfamidates for asymmetric addition of butyllithium to benzaldehyde.
Organic letters (2007-08-19)
Petra Rönnholm, Mikael Södergren, Göran Hilmersson
ABSTRACT
A robust and scaleable route to chiral 1-isopropylamino-2-(diphenylphosphino)ethanes is described via the ring-opening of chiral, cyclic sulfamidates with potassium diphenylphosphide (KPPh(2)). The novel protocol offers a robust access to gram quantities of chiral amino phosphinoethanes in high yields. The Li-amides of the chiral aminophosphines were evaluated as chiral ligands in the asymmetric addition of n-butyllithium (BuLi) to benzaldehyde, yielding 1-phenylpentanol up to 98% ee.
Sigma-Aldrich
n-Butyllithium solution, 2.7 M in heptane
Sigma-Aldrich
n-Butyllithium solution, 2.5 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 2.0 M in cyclohexane
Sigma-Aldrich
n-Butyllithium solution, 1.6 M in hexanes
Sigma-Aldrich
n-Butyllithium solution, 11.0 M in hexanes