Merck
  • Combined liquid chromatography-tandem mass spectrometry as an analytical method for high throughput screening for X-linked adrenoleukodystrophy and other peroxisomal disorders: preliminary findings.
Combined liquid chromatography-tandem mass spectrometry as an analytical method for high throughput screening for X-linked adrenoleukodystrophy and other peroxisomal disorders: preliminary findings.
Molecular genetics and metabolism (2006-07-11)
Walter C Hubbard, Ann B Moser, Silvia Tortorelli, Anita Liu, David Jones, Hugo Moser
ABSTRACT
Utilizing combined liquid chromatography-tandem mass spectrometry (LC-MS/MS) as the analytical method, we have demonstrated a ten to sixtyfold excess of lysophosphatidyl choline containing hexacosanoic acid (26:0) in dried blood spots on a filter paper matrix from 25 male patients with X-linked adrenoleukodystrophy and nine patients with peroxisome biogenesis disorders compared to 19 controls. There was no overlap between normal subjects versus affected subjects.
Sigma-Aldrich
Hexacosanoic acid, ≥95% (capillary GC)
Sigma-Aldrich
Hexacosanoic acid, technical, ≥90% (GC)
Avanti
22:0 Lyso PC, 1-behenoyl-2-hydroxy-sn-glycero-3-phosphocholine, powder