Merck
Behavioral monitoring of trained insects for chemical detection.
Biotechnology progress (2006-02-04)
Glen C Rains, Samuel L Utley, W Joe Lewis
ABSTRACT
A portable, handheld volatile odor detector ("Wasp Hound") that utilizes a computer vision system and Microplitis croceipes (Cresson) (Hymenoptera: Braconidae), a parasitoid wasp, as the chemical sensor was created. Five wasps were placed in a test cartridge and placed inside the device. Wasps were either untrained or trained by associative learning to detect 3-octanone, a common fungal volatile chemical. The Wasp Hound sampled air from the headspace of corn samples prepared within the lab and, coupled with Visual Cortex, a software program developed using the LabView graphical programming language, monitored and analyzed wasp behavior. The Wasp Hound, with conditioned wasps, was able to detect 0.5 mg of 3-octanone within a 240 mL glass container filled with feed corn ( approximately 2.6 x 10(-5) mol/L). The Wasp Hound response to the control (corn alone) and a different chemical placed in the corn (0.5 mg of myrcene) was significantly different than the response to the 3-octanone. Wasp Hound results from untrained wasps were significantly different from trained wasps when comparing the responses to 3-octanone. The Wasp Hound may provide a unique method for monitoring grains, peanuts, and tree nuts for fungal growth associated with toxin production, as well as detecting chemicals associated with forensic investigations and plant/animal disease.
Sigma-Aldrich
3-Octanone, ≥98%, FG
Supelco
3-Octanone, analytical standard
Sigma-Aldrich
3-Octanone, ≥98%