Merck
  • Towards evidence based emergency medicine: best BETs from the Manchester Royal Infirmary. Phenytoin or paraldehyde as the second drug for convulsions in children.
Towards evidence based emergency medicine: best BETs from the Manchester Royal Infirmary. Phenytoin or paraldehyde as the second drug for convulsions in children.
Emergency medicine journal : EMJ (2002-01-05)
Will Townend, Kevin Mackway-Jones
ABSTRACT
A short cut review was carried out to establish whether phenytoin or paraldehyde should be given as the second drug for resistant fits in children. Altogether 41 papers were found using the reported search, of which none answered the question posed. Further research is needed in this area.
Sigma-Aldrich
Paraldehyde, ≥97.0% (GC)
Sigma-Aldrich
Paraldehyde, ≥97%
Sigma-Aldrich
Paraldehyde, Arxada quality, ≥99.5% (GC)