Merck
  • Aedes aegypti (Diptera: Culicidae) age determination by cuticular hydrocarbon analysis of female legs.
Aedes aegypti (Diptera: Culicidae) age determination by cuticular hydrocarbon analysis of female legs.
Journal of medical entomology (1999-12-11)
M L Desena, J D Edman, J M Clark, S B Symington, T W Scott
ABSTRACT
We previously described methods for age-grading female Aedes aegypti (L.) by gas chromatographic (GC) analysis of whole-body cuticular hydrocarbon patterns. Two regression models were developed that were based on the age-dependent, relative abundance of 2 cuticular hydrocarbons, pentacosane (GC peak 1) and nonacosane (GC peak 5). We have refined this method so that only the legs are required to age individual females. Two new regression models were developed that also use the relative abundance of a 3rd cuticular hydrocarbon, octacosane (GC peak 4). These models improve the overall accuracy of the cuticular hydrocarbon method for aging female mosquitoes, especially for older females from 132 to 165 degree-days (DD) of age (12-15 calendar days at 28 degrees C). The correlation coefficients (R2) for the best-fitted linear regression models for aging females from 0 to 132 and 0-165 DD were 0.80 and 0.81, respectively (P < 0.001 in all cases). The use of leg cuticular hydrocarbons for estimating the age of female Ae. aegypti has a significant advantage over whole-body extracts as indicated by the decreased variability associated with the relative abundance of pentacosane and the expanded range over which the models were able to predict age accurately by the addition of the relative abundance of octacosane.
Sigma-Aldrich
Nonacosane, 99%
Supelco
Nonacosane, analytical standard