Merck
  • Rosuvastatin inhibits TGF-beta1 expression and alleviates myocardial fibrosis in diabetic rats.
Rosuvastatin inhibits TGF-beta1 expression and alleviates myocardial fibrosis in diabetic rats.
Die Pharmazie (2013-06-28)
Yu-Xiao Ma, Wei-Hua Li, Qiang Xie
ABSTRACT
This study aimed to investigate the effects of rosuvastatin on TGF-beta1 expression, cardiac fibrosis, ventricular remodeling and cardiac function in diabetic cardiomyopathy rats. Twenty-seven diabetic rats induced by streptozotocin intraperitoneal injection were randomly divided into three groups, viz. diabetic, rosuvastatin low-dose (Ros-L) and high dose group (Ros-H). Intervention group were given rosuvastatin 2 mg/kg/d and 5 mg/kg/d orally, respectively. After 10 weeks, the levels of glycosylated hemoglobin (HbA1c), creatine phosphokinase isoenzyme (CK-MB), plasma brain natriuretic peptide (BNP), myocardial collagen volume fraction (CVF) and left ventricular mass index (LVWI) were measured. CK-MB levels in Ros-H and Ros-L rats were lower than in the diabetic group. Rosuvastatin alleviated myofibrosis cordis and fibroplastic proliferation. LVWI, BNP, CVF and TGF-beta1 mRNA and protein levels in the diabetic group were higher than in the control, but were reduced after rosuvastatin treatment. These results demonstrate that rosuvastatin dose-dependently reduces TGF-beta1 expression and inhibits the development of myocardial fibrosis in diabetic cardiomyopathy.
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type III (Sigma Type X), powder
Sigma-Aldrich
Creatine Phosphokinase from bovine heart, Type III, salt-free, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Collagen from bovine nasal septum, Bornstein and Traub Type II, powder
Sigma-Aldrich
Collagen from bovine tracheal cartilage, Bornstein and Traub Type II, powder
Sigma-Aldrich
Creatine Phosphokinase from rabbit muscle, Type I, salt-free, lyophilized powder, ≥150 units/mg protein
Sigma-Aldrich
Collagen from rabbit skin, Bornstein and Traub Type I, powder
Sigma-Aldrich
Collagen from Engelbreth-Holm-Swarm murine sarcoma basement membrane, Type IV (Miller), lyophilized powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from rat tail, Bornstein and Traub Type I, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen, Type I solution from rat tail, BioReagent, suitable for cell culture, sterile-filtered
Sigma-Aldrich
Collagen Type IV from human cell culture, Bornstein and Traub Type IV, 0.3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen human, Bornstein and Traub Type I, acid soluble, powder, ~95% (SDS-PAGE)
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type I (Sigma Type VIII), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type V (Sigma Type IX), powder
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from calf skin, Bornstein and Traub Type I, (0.1% solution in 0.1 M acetic acid), aseptically processed, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, powder
Sigma-Aldrich
Collagen from bovine achilles tendon, powder, suitable for substrate for collagenase
Sigma-Aldrich
Collagen from chicken sternal cartilage, Type II (Miller), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Collagen from human placenta, Bornstein and Traub Type IV, solution, suitable for cell culture, High Performance