Merck
Accumulation of senescent cells in the stroma of aged mouse ovary.
The Journal of reproduction and development (2023-11-06)
Natsumi Maruyama, Isuzu Fukunaga, Tomoaki Kogo, Tsutomu Endo, Wataru Fujii, Masami Kanai-Azuma, Kunihiko Naito, Koji Sugiura
ABSTRACT
Senescent cells play a detrimental role in age-associated pathogenesis by producing factors involved in senescence-associated secretory phenotype (SASP). The present study was conducted to examine the possibility that senescent cells are present in aged ovaries and, if so, to determine the tissue region where senescent cells accumulate using a mouse model. Female mice at 2-4 and 8-10 months were used as reproductively young and aged models, respectively; the latter included mice with and without reproductive experience. Cells positive for senescence-associated β-galactosidase (SA-β-Gal) staining, one of the markers of cellular senescence, were detected in the stromal region of aged, but not young, ovaries regardless of reproductive experience. Likewise, the localization of cells expressing CDKN2A (cyclin dependent kinase inhibitor 2A), another senescence marker, in the stromal region of aged ovaries was detected with immunohistochemistry. CDKN2A expression detected by western blotting was significantly higher in the ovaries of aged mice with reproductive experience than in those without the experience. Moreover, cells positive for both γH2AX (a senescence marker) and fluorescent SA-β-Gal staining were present in those isolated from aged ovaries. In addition, the transcript levels of several SASP factors were significantly increased in aged ovaries. These results suggest that senescent cells accumulate in the ovarian stroma and may affect ovarian function in aged mice. Additionally, reproductive experience may promote accumulation.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Millipore
Goat Anti-Mouse IgG Antibody, Rhodamine conjugate, Species Adsorbed, 1.4 mg/mL, Chemicon®
Sigma-Aldrich
Goat Anti-Rabbit IgG Antibody, Peroxidase Conjugated, 1 mg/mL (after reconstitution), Chemicon®