Merck
  • A Whole-Genome Sequencing Study Implicates GRAMD1B in Multiple Sclerosis Susceptibility.
A Whole-Genome Sequencing Study Implicates GRAMD1B in Multiple Sclerosis Susceptibility.
Genes (2022-12-24)
Federica Esposito, Ana Maria Osiceanu, Melissa Sorosina, Linda Ottoboni, Bryan Bollman, Silvia Santoro, Barbara Bettegazzi, Andrea Zauli, Ferdinando Clarelli, Elisabetta Mascia, Andrea Calabria, Daniele Zacchetti, Ruggero Capra, Maurizio Ferrari, Paolo Provero, Dejan Lazarevic, Davide Cittaro, Paola Carrera, Nikolaos Patsopoulos, Daniela Toniolo, A Dessa Sadovnick, Gianvito Martino, Philip L De Jager, Giancarlo Comi, Elia Stupka, Carles Vilariño-Güell, Laura Piccio, Filippo Martinelli Boneschi
ABSTRACT
While the role of common genetic variants in multiple sclerosis (MS) has been elucidated in large genome-wide association studies, the contribution of rare variants to the disease remains unclear. Herein, a whole-genome sequencing study in four affected and four healthy relatives of a consanguineous Italian family identified a novel missense c.1801T > C (p.S601P) variant in the GRAMD1B gene that is shared within MS cases and resides under a linkage peak (LOD: 2.194). Sequencing GRAMD1B in 91 familial MS cases revealed two additional rare missense and two splice-site variants, two of which (rs755488531 and rs769527838) were not found in 1000 Italian healthy controls. Functional studies demonstrated that GRAMD1B, a gene with unknown function in the central nervous system (CNS), is expressed by several cell types, including astrocytes, microglia and neurons as well as by peripheral monocytes and macrophages. Notably, GRAMD1B was downregulated in vessel-associated astrocytes of active MS lesions in autopsied brains and by inflammatory stimuli in peripheral monocytes, suggesting a possible role in the modulation of inflammatory response and disease pathophysiology.
Sigma-Aldrich
Anti-GRAMD1B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Interferon-γ human, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), lyophilized powder, suitable for cell culture