Merck
  • MicroRNA21 inhibition affects porcine oocyte maturation and alters protein expression critical for metabolic pathway function.
MicroRNA21 inhibition affects porcine oocyte maturation and alters protein expression critical for metabolic pathway function.
Molecular reproduction and development (2022-08-25)
Yunsheng Li, Malavika K Adur, Steven M Lonergan, Aileen F Keating, Jason W Ross
ABSTRACT
MicroRNA21 (MIR21) abundance in porcine oocytes and cumulus cells increases during in vitro maturation. The mechanism by which MIR21 regulates oocyte maturation and the effect on the developmental competence of subsequent embryos remains unclear. The objective of this study was to assess the function of MIR21 during porcine oocyte maturation and its effect on embryonic development. Treatment with peptide nucleic acid MIR21 inhibitor (MIR21-PNA), designed to specifically bind to and prevent MIR21 activity during in vitro oocyte maturation, decreased cumulus cell expansion, and the oocyte ability to achieve metaphase II maturation stage when compared to control groups. Following parthenogenetic activation, the cleavage rate at 48 h in the MIR21-PNA group was decreased (p ≤ 0.03) relative to the control groups. Additionally, liquid chromatography-mass spectrometry (LC-MS/MS) of oocyte and cumulus cell total protein following MIR21-PNA treatment during in vitro maturation identified changes in signaling pathways with primary involvement of glucose metabolism (GM) pathways. Furthermore, there was no difference (p = 0.21) in oocyte maturation of control and MIR21-PNA treated oocytes when cultured in pyruvate lacking medium. Finally, MIR21-PNA treatment decreased (p = 0.04) glutathione and increased (p = 0.07) reactive oxygen species production in the oocyte. These data suggest that MIR21 influences porcine oocyte maturation by regulating GM pathways in the cumulus-oocyte complex.
Sigma-Aldrich
Anti-GCLC antibody produced in rabbit, affinity isolated antibody