Merck
  • Differential role of Pax6 and its interaction with Shh-Gli1-IDH2 axis in regulation of glioma growth and chemoresistance.
Differential role of Pax6 and its interaction with Shh-Gli1-IDH2 axis in regulation of glioma growth and chemoresistance.
Journal of biochemical and molecular toxicology (2022-10-08)
Shirin Farheen, Swalih P Ahmed, Mubeena Mariyath P M, Tasneem Kausar, Md Fakhrul Hoda, Sayeedul H Arif, Shahid M Nayeem, Asif Ali, Kunzang Chosdol, Mehdi H Shahi
ABSTRACT
Glioma is a major brain tumor, and the associated mortality rate is very high. Contemporary therapies provide a chance of survival for 9-12 months. Therefore, a novel approach is essential to improve the survival rate. Sonic hedgehog (Shh) cell signaling is critical for early development in various tumors. This investigation attempted to explore the potential interaction and regulation of Shh-Gli1 cell signaling in association with paired box 6 (Pax6) and isocitrate dehydrogenase 2 (IDH2). The expression pattern of Shh, Gli1, Pax6, and IDH2 was examined by transcriptome analysis, immunohistochemistry, and confocal images. The results suggest the interaction of Shh-Gli1 cell signaling pathway with Pax6 and IDH2 and potential regulation. Thereafter, we performed protein-protein docking and molecular dynamic simulations (MDS) of Gli1 with Pax6 and IDH2. The results suggest differential dynamic interactions of Gli1-IDH2 and Gli1-Pax6. Gli1 knockdown downregulated the expression of Pax6 and upregulated the expression of IDH2. Moreover, Gli1 knockdown decreased the expression of the drug resistance gene MRP1. The knockdown of Pax6 gene in glioma cells downregulated the expression of Gli1 and IDH2 and promoted cell proliferation. Moreover, the efficacy of the treatment of glioma cells with temozolomide (TMZ) and Gli1 inhibitor GANT61 was higher than that of TMZ alone. MDS results revealed that the interactions of Gli1 with IDH2 were stronger and more stable than those with Pax6. Intriguingly, inhibition of Pax6 promoted glioma growth even in the presence of TMZ. However, the tumor-suppressive nature of Pax6 was altered when Gli1 was inhibited by GANT61, and it showed potential oncogenic character, as observed in other cancers. Therefore, we conclude that Pax6 interacted with IDH2 and Gli1 in glioma. Moreover, the Shh-Gli1-IDH2/Pax6 cell signaling axis provides a new therapeutic approach for inhibiting the progression of the disease and mitigating drug resistance in glioma.
Sigma-Aldrich
SAG dihydrochloride, ≥98% (HPLC)
Sigma-Aldrich
GANT61, ≥98% (HPLC)