Merck
  • Enzymatic control of chitosan gelation for delivery of periodontal ligament cells.
Enzymatic control of chitosan gelation for delivery of periodontal ligament cells.
Macromolecular bioscience (2014-04-02)
Xiang-Zhen Yan, Arnold W G Nijhuis, Jeroen J J P van den Beucken, Sanne K Both, John A Jansen, Sander C G Leeuwenburgh, Fang Yang
ABSTRACT
The aim of this study is to optimize enzymatic control over gelation of chitosan-based hydrogels for the delivery of periodontal ligament cells (PDLCs). The results reveal that the gelation time, strength, and degradation rate of the chitosan hydrogels can be controlled precisely by variation of the urea and urease concentrations. PDLCs remain viable inside these hydrogels for up to 30 days. Cells released from the hydrogel upon degradation and collected after 3, 15, and 30 days are able to form colonies and osteogenically differentiate. In conclusion, the enzymatic control over the gelation of chitosan hydrogels offers options for the delivery of PDLCs.
Sigma-Aldrich
Urease from Canavalia ensiformis (Jack bean), Type III, powder, 15,000-50,000 units/g solid