Merck
  • Characterization of cerebrospinal fluid biomarkers associated with neurodegenerative diseases in healthy cynomolgus and rhesus macaque monkeys.
Characterization of cerebrospinal fluid biomarkers associated with neurodegenerative diseases in healthy cynomolgus and rhesus macaque monkeys.
Alzheimer's & dementia (New York, N. Y.) (2022-04-14)
Emma L Robertson, Susan E Boehnke, Natalia de M Lyra E Silva, Brittney Armitage-Brown, Andrew Winterborn, Douglas J Cook, Fernanda G De Felice, Douglas P Munoz
ABSTRACT
Monkeys are becoming important translational models of neurodegenerative disease. To facilitate model development, we measured cerebrospinal fluid (CSF) concentrations of key biomarkers in healthy male and female cynomolgus and rhesus macaques. Amyloid beta (Aβ40, Aβ42), tau (total tau [t-tau], phosphorylated tau [pThr181]), and neurofilament light (NfL) concentrations were measured in CSF of 82 laboratory-housed, experimentally naïve cynomolgus (n = 33) and rhesus (n = 49) macaques. Aβ40 and Aβ42 were significantly higher in rhesus, and female rhesus were higher than males. NfL and t-tau were higher in males, and NfL was higher in rhesus macaques. p-tau was not affected by species or sex. We also examined whether sample location (lumbar or cisterna puncture) affected concentrations. Sample acquisition site only affected NfL, which was higher in CSF from lumbar puncture compared to cisterna magna puncture. Establishing normative biomarker values for laboratory-housed macaque monkeys provides an important resource by which to compare to monkey models of neurodegenerative diseases.
Product Description
Millipore
MILLIPLEX® Human Amyloid Beta and Tau Magnetic Bead Panel - Multiplex Assay, allows quantitative multiplex detection of multiple analytes simultaneously