Merck
  • SIRT4 is an early regulator of branched-chain amino acid catabolism that promotes adipogenesis.
SIRT4 is an early regulator of branched-chain amino acid catabolism that promotes adipogenesis.
Cell reports (2021-07-15)
Elma Zaganjor, Haejin Yoon, Jessica B Spinelli, Elizabeth R Nunn, Gaëlle Laurent, Paulina Keskinidis, Suganja Sivaloganathan, Shakchhi Joshi, Giulia Notarangelo, Stacy Mulei, Mathew T Chvasta, Sarah A Tucker, Krystle Kalafut, Robert A H van de Ven, Clary B Clish, Marcia C Haigis
ABSTRACT
Upon nutrient stimulation, pre-adipocytes undergo differentiation to transform into mature adipocytes capable of storing nutrients as fat. We profiled cellular metabolite consumption to identify early metabolic drivers of adipocyte differentiation. We find that adipocyte differentiation raises the uptake and consumption of numerous amino acids. In particular, branched-chain amino acid (BCAA) catabolism precedes and promotes peroxisome proliferator-activated receptor gamma (PPARγ), a key regulator of adipogenesis. In early adipogenesis, the mitochondrial sirtuin SIRT4 elevates BCAA catabolism through the activation of methylcrotonyl-coenzyme A (CoA) carboxylase (MCCC). MCCC supports leucine oxidation by catalyzing the carboxylation of 3-methylcrotonyl-CoA to 3-methylglutaconyl-CoA. Sirtuin 4 (SIRT4) expression is decreased in adipose tissue of numerous diabetic mouse models, and its expression is most correlated with BCAA enzymes, suggesting a potential role for SIRT4 in adipose pathology through the alteration of BCAA metabolism. In summary, this work provides a temporal analysis of adipocyte differentiation and uncovers early metabolic events that stimulate transcriptional reprogramming.
Sigma-Aldrich
Anti-Actin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Insulin solution from bovine pancreas, 10 mg/mL insulin in 25  mM HEPES, pH 8.2, BioReagent, sterile-filtered, suitable for cell culture
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, BioUltra, ≥99%
Sigma-Aldrich
β-Methylcrotonyl coenzyme A lithium salt, ≥90%
Sigma-Aldrich
Anti-methylglutaryl-lysine, from rabbit