Merck
  • High refractive index monofunctional monomers as promising diluents for dental composites.
High refractive index monofunctional monomers as promising diluents for dental composites.
Dental materials : official publication of the Academy of Dental Materials (2020-12-29)
Yohann Catel, Jörg Angermann, Pascal Fässler, Urs Fischer, Thomas Schnur, Norbert Moszner
ABSTRACT
To evaluate high refractive index methacrylates as diluents for the formulation of radiopaque esthetic bulk-fill composites. 2-(4-Cumylphenoxy)ethyl methacrylate 1, 2-(2-phenylphenoxy)ethyl methacrylate 2 and 2-[2-(2-phenylphenoxy)ethoxy]ethyl methacrylate 3 were synthesized and characterized by 1H NMR spectroscopy. The reactivity of each monomer was studied using photo-DSC. Bulk-fill composites based on monomers 1-3 were formulated. Translucency (before and after light cure) was measured using a spectrophotometer. The depth of cure and the water sorption of these materials were determined according to ISO 4049. The flexural strength and modulus of elasticity were measured using a three-point bending setup, according to ISO 4049. The shrinkage force was assessed based on a method described by Watts et al. using a universal testing machine. Monomers 1-3 were easily synthesized in two steps. They exhibit a low viscosity and a high refractive index (1.553-1.573). Monofunctional methacrylates 1-3 were found to be more reactive than triethylene glycol dimethacrylate (TEGDMA). Bulk-fill composites based on these monomers were successfully prepared. They exhibit a high depth of cure and excellent esthetic properties (low transparency). These composites provide higher flexural modulus as well as lower water sorption than a corresponding material based on TEGDMA. Methacrylates 1 and 3 are particularly interesting as they led to composites showing lower shrinkage force. Methacrylates 1-3 are promising diluents for the formulation of highly esthetic radiopaque bulk-fill composites.
Sigma-Aldrich
4-Cumylphenol, 99%
Sigma-Aldrich
Benzyltributylammonium chloride, ≥98%
Sigma-Aldrich
2-(2-Chloroethoxy)ethanol, 99%
Sigma-Aldrich
Methacrylic anhydride, contains 2,000 ppm topanol A as inhibitor, ≥98%