Merck
  • Tacripyrines, the first tacrine-dihydropyridine hybrids, as multitarget-directed ligands for the treatment of Alzheimer's disease.
Tacripyrines, the first tacrine-dihydropyridine hybrids, as multitarget-directed ligands for the treatment of Alzheimer's disease.
Journal of medicinal chemistry (2009-04-21)
José Marco-Contelles, Rafael León, Cristóbal de los Ríos, Abdelouahid Samadi, Manuela Bartolini, Vincenza Andrisano, Oscar Huertas, Xavier Barril, F Javier Luque, María I Rodríguez-Franco, Beatriz López, Manuela G López, Antonio G García, María do Carmo Carreiras, Mercedes Villarroya
ABSTRACT
Tacripyrines (1-14) have been designed by combining an AChE inhibitor (tacrine) with a calcium antagonist such as nimodipine and are targeted to develop a multitarget therapeutic strategy to confront AD. Tacripyrines are selective and potent AChE inhibitors in the nanomolar range. The mixed type inhibition of hAChE activity of compound 11 (IC(50) 105 +/- 15 nM) is associated to a 30.7 +/- 8.6% inhibition of the proaggregating action of AChE on the Abeta and a moderate inhibition of Abeta self-aggregation (34.9 +/- 5.4%). Molecular modeling indicates that binding of compound 11 to the AChE PAS mainly involves the (R)-11 enantiomer, which also agrees with the noncompetitive inhibition mechanism exhibited by p-methoxytacripyrine 11. Tacripyrines are neuroprotective agents, show moderate Ca(2+) channel blocking effect, and cross the blood-brain barrier, emerging as lead candidates for treating AD.
Product Description
Supelco
Melting point standard 235-237°C, analytical standard
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Progesterone, meets USP testing specifications
Sigma-Aldrich
Progesterone, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Atenolol, ≥98% (TLC), powder
Sigma-Aldrich
Hydrocortisone-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Supelco
Testosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Progesterone, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Progesterone, ≥99%
Supelco
Caffeine solution, analytical standard, 1.0 mg/mL in methanol
Supelco
Progesterone, VETRANAL®, analytical standard
Supelco
Resveratrol, analytical standard
Supelco
Ofloxacin, VETRANAL®, analytical standard
Supelco
Mettler-Toledo Calibration substance ME 18872, Caffeine, traceable to primary standards (LGC)
Sigma-Aldrich
Caffeine, anhydrous, tested according to Ph. Eur.
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Caffeine, Sigma Reference Standard, vial of 250 mg
Sigma-Aldrich
Caffeine, meets USP testing specifications, anhydrous
Sigma-Aldrich
Caffeine, BioXtra
Sigma-Aldrich
Piroxicam, meets USP testing specifications
Sigma-Aldrich
Progesterone-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrocortisone solution, 50 μM, sterile-filtered, BioXtra, suitable for cell culture
Supelco
Testosterone solution, 1.0 mg/mL in 1,2-dimethoxyethane, drug standard