Merck
  • Proteasome-Bound UCH37/UCHL5 Debranches Ubiquitin Chains to Promote Degradation.
Proteasome-Bound UCH37/UCHL5 Debranches Ubiquitin Chains to Promote Degradation.
Molecular cell (2020-11-07)
Kirandeep K Deol, Sean O Crowe, Jiale Du, Heather A Bisbee, Robert G Guenette, Eric R Strieter, Kirandeep K Deol, Sean O Crowe, Jiale Du, Heather A Bisbee, Robert G Guenette, Eric R Strieter
ABSTRACT
The linkage, length, and architecture of ubiquitin (Ub) chains are all important variables in providing tight control over many biological paradigms. There are clear roles for branched architectures in regulating proteasome-mediated degradation, but the proteins that selectively recognize and process these atypical chains are unknown. Here, using synthetic and enzyme-derived ubiquitin chains along with intact mass spectrometry, we report that UCH37/UCHL5, a proteasome-associated deubiquitinase, cleaves K48 branched chains. The activity and selectivity toward branched chains is markedly enhanced by the proteasomal Ub receptor RPN13/ADRM1. Using reconstituted proteasome complexes, we find that chain debranching promotes degradation of substrates modified with branched chains under multi-turnover conditions. These results are further supported by proteome-wide pulse-chase experiments, which show that the loss of UCH37 activity impairs global protein turnover. Our work therefore defines UCH37 as a debranching deubiquitinase important for promoting proteasomal degradation.
Sigma-Aldrich
Doxycycline hyclate
Sigma-Aldrich
DBCO-Cy5, for Copper-free Click Chemistry
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Maleimide, 99%
Sigma-Aldrich
Formic acid, ACS reagent, ≥88%