Merck
  • Proline-rich protein PRR19 functions with cyclin-like CNTD1 to promote meiotic crossing over in mouse.
Proline-rich protein PRR19 functions with cyclin-like CNTD1 to promote meiotic crossing over in mouse.
Nature communications (2020-06-20)
Anastasiia Bondarieva, Kavya Raveendran, Vladyslav Telychko, H B D Prasada Rao, Ramya Ravindranathan, Chrysoula Zorzompokou, Friederike Finsterbusch, Ihsan Dereli, Frantzeskos Papanikos, Daniel Tränkner, Alexander Schleiffer, Ji-Feng Fei, Anna Klimova, Masaru Ito, Dhananjaya S Kulkarni, Ingo Roeder, Neil Hunter, Attila Tóth
ABSTRACT
Orderly chromosome segregation is enabled by crossovers between homologous chromosomes in the first meiotic division. Crossovers arise from recombination-mediated repair of programmed DNA double-strand breaks (DSBs). Multiple DSBs initiate recombination, and most are repaired without crossover formation, although one or more generate crossovers on each chromosome. Although the underlying mechanisms are ill-defined, the differentiation and maturation of crossover-specific recombination intermediates requires the cyclin-like CNTD1. Here, we identify PRR19 as a partner of CNTD1. We find that, like CNTD1, PRR19 is required for timely DSB repair and the formation of crossover-specific recombination complexes. PRR19 and CNTD1 co-localise at crossover sites, physically interact, and are interdependent for accumulation, indicating a PRR19-CNTD1 partnership in crossing over. Further, we show that CNTD1 interacts with a cyclin-dependent kinase, CDK2, which also accumulates in crossover-specific recombination complexes. Thus, the PRR19-CNTD1 complex may enable crossover differentiation by regulating CDK2.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, for general use, Type I, ≥125 CDU/mg solid
Sigma-Aldrich
Immobilon®-P Polyvinylidene difluoride membranes, size 26.5 cm × 375 cm
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 1, DMSO solution
Sigma-Aldrich
Phosphatase Inhibitor Cocktail 2, aqueous solution (dark coloration may develop upon storage, which does not affect the activity)
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Sudan Black B, certified by the Biological Stain Commission
Sigma-Aldrich
Tyrode′s Solution, Acidic, liquid, sterile-filtered, suitable for mouse embryo cell culture
Sigma-Aldrich
Milrinone, ≥97% (TLC), powder
Sigma-Aldrich
M2 medium, With HEPES, without penicillin and streptomycin, liquid, sterile-filtered, suitable for mouse embryo cell culture
Sigma-Aldrich
Spermidine, ≥99% (GC)
Sigma-Aldrich
Anti-α-Tubulin antibody, Mouse monoclonal, clone DM1A, purified from hybridoma cell culture