Merck
Structural basis of lectin-carbohydrate recognition.
Annual review of biochemistry (1996-01-01)
W I Weis, K Drickamer
ABSTRACT
Lectins are responsible for cell surface sugar recognition in bacteria, animals, and plants. Examples include bacterial toxins; animal receptors that mediate cell-cell interactions, uptake of glycoconjugates, and pathogen neutralization; and plant toxins and mitogens. The structural basis for selective sugar recognition by members of all of these groups has been investigated by x-ray crystallography. Mechanisms for sugar recognition have evolved independently in diverse protein structural frameworks, but share some key features. Relatively low affinity binding sites for monosaccharides are formed at shallow indentations on protein surfaces. Selectivity is achieved through a combination of hydrogen bonding to the sugar hydroxyl groups with van der Waals packing, often including packing of a hydrophobic sugar face against aromatic amino acid side chains. Higher selectivity of binding is achieved by extending binding sites through additional direct and water-mediated contacts between oligosaccharides and the protein surface. Dramatically increased affinity for oligosaccharides results from clustering of simple binding sites in oligomers of the lectin polypeptides. The geometry of such oligomers helps to establish the ability of the lectins to distinguish surface arrays of polysaccharides in some instances and to crosslink glycoconjugates in others.
Product Description
Avanti
C12-NBD Lactosyl Ceramide, Avanti Researchā„¢ - A Croda Brand 810227P, powder
Avanti
C6-NBD Lactosyl Ceramide, Avanti Researchā„¢ - A Croda Brand 810226P, powder