Merck
  • Cell-based, animal and H1 receptor binding studies relative to the sedative effects of ketotifen and norketotifen atropisomers.
Cell-based, animal and H1 receptor binding studies relative to the sedative effects of ketotifen and norketotifen atropisomers.
The Journal of pharmacy and pharmacology (2020-02-08)
Feifei Feng, J Paul Fawcett, Hu Zhang, Ian G Tucker
ABSTRACT
Ketotifen (K) and its active metabolite norketotifen (N) exist as optically active atropisomers. They both have antihistaminic and anti-inflammatory properties but the S-atropisomer of N (SN) causes less sedation than K and RN in rodents. This study investigated whether this could be related to a lower concentration of SN in brain or a lower affinity of SN for rat brain H1 receptors. Ketotifen and norketotifen atropisomers were quantified using a validated chiral HPLC assay. RBE4 and Caco-2 cell monolayers were used in uptake and permeability studies, respectively. Free and total brain-to-plasma (B/P) ratios were determined after injecting racemic K and N into rat tail veins. Affinity for rat brain H1 receptors (KI ) was determined using the [3 H]mepyramine binding assay. Uptake and permeation studies indicate no stereoselective transport for K or N. B/P ratios reveal the brain concentration of N is lower than K with no stereoselective transport into brain. Finally, the [3 H]mepyramine binding assay shows SN has the lowest affinity for rat brain H1 receptors. The lower sedative effect of SN in rodents is probably due to a combination of a lower uptake of N than K into the brain and less affinity of SN for CNS H1 receptors.
Sigma-Aldrich
Calcium chloride, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Triprolidine hydrochloride, ≥99%