Merck
  • Transmembrane Protein 39A Promotes the Replication of Encephalomyocarditis Virus via Autophagy Pathway.
Transmembrane Protein 39A Promotes the Replication of Encephalomyocarditis Virus via Autophagy Pathway.
Frontiers in microbiology (2019-12-19)
Xiangrong Li, Ruixian Ma, Qian Li, Shengjun Li, Haixia Zhang, Jingying Xie, Jialin Bai, Adi Idris, Ruofei Feng
ABSTRACT
Encephalomyocarditis virus (EMCV) causes encephalitis, myocarditis, neuropathy, reproductive disorders, and diabetes in animals. EMCV is known to induce cell autophagy; however, the molecular mechanisms underlying this remain unclear. Here, we show that the type III-transmembrane protein, transmembrane protein 39A (TMEM39A), plays a critical role in EMCV replication. We showed that EMCV GS01 strain infection upregulated TMEM39A expression. Importantly, EMCV induced autophagy in a range of host cells. The autophagy chemical inhibitor, 3-MA, inhibited EMCV replication and reduced TMEM39A expression. This is the first study demonstrating TMEM39A promoting the replication of EMCV via autophagy. Overall, we show that TMEM39A plays a positive regulatory role in EMCV proliferation and that TMEM39A expression is dependent on the autophagy pathway.
Product Description
Millipore
Immobilon®-PSQ PVDF Membrane, 1 roll, 27 cm x 3.75 m, 0.2 µm pore size, Hydrophobic PVDF Transfer Membrane for western blotting.
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor